Learn More
Chemglass Life Sciences Chemglass Cryogenic Vials, 1.9 mL, 3D Barcode, External Thread, Caps On, Racked, Sterile
SDP
Catalog No. 502733352
Encompass_Preferred
Chemglass 1.9 mL cryogenic vials designed for ultra-low temperature sample storage and automated handling. The vials include a three-code-in-one identification system for reliable sample tracking and are supplied sterile in SBS-format racks for high-throughput workflows.
  • Capacity: 1.9 mL.
  • Identification: 3D barcode system with 2D code on the bottom and 1D code on the side.
  • Thread: external threaded vial.
  • Cap: caps on configuration for secure closure.
  • Packaging: racked and sterile, 48 per pack, 480 per case.
  • Compatibility: SBS-format racks compatible with common automated systems.
  • Material and purity: medical-grade polypropylene, DNase/RNase free, non-endotoxin.
  • Standards: meets USP Class VI biocompatibility standards.
Catalog No. 50-273-3352 Supplier Chemglass Life Sciences Supplier No. CGN476503
Chemglass 1.9 mL cryogenic vials designed for ultra-low temperature sample storage and automated handling. The vials include a three-code-in-one identification system for reliable sample tracking and are supplied sterile in SBS-format racks for high-throughput workflows.
  • Capacity: 1.9 mL.
  • Identification: 3D barcode system with 2D code on the bottom and 1D code on the side.
  • Thread: external threaded vial.
  • Cap: caps on configuration for secure closure.
  • Packaging: racked and sterile, 48 per pack, 480 per case.
  • Compatibility: SBS-format racks compatible with common automated systems.
  • Material and purity: medical-grade polypropylene, DNase/RNase free, non-endotoxin.
  • Standards: meets USP Class VI biocompatibility standards.