Learn More
Chemglass Life Sciences Chemglass Cryogenic Vials, 1.9 mL, 3D Barcode, SBS Format, External Thread, Sterile (CGN-4765-05)
SDP
Catalog No. 502733350
Encompass_Preferred
Chemglass Cryogenic Vials (CGN-4765-05) are 1.9 mL medical-grade polypropylene vials with integrated multi-code identification designed for long-term cryogenic storage and automated sample handling. They are sterile and compatible with SBS-format automation and common cryo racks.
  • 1.9 mL capacity.
  • Medical-grade cryogenic polypropylene (USP Class-6).
  • Three-code-in-one identification: 2D bottom barcode and 1D side barcode.
  • SBS-format external thread; caps supplied separated.
  • Electron beam sterilized, SAL = 10^-6.
  • Compatible with Brooks System and NEST racks.
  • Suitable for use at liquid nitrogen temperatures (-196 °C).
  • Packaged 48 per pack, 480 per case.
Catalog No. 50-273-3350 Supplier Chemglass Life Sciences Supplier No. CGN476505
Chemglass Cryogenic Vials (CGN-4765-05) are 1.9 mL medical-grade polypropylene vials with integrated multi-code identification designed for long-term cryogenic storage and automated sample handling. They are sterile and compatible with SBS-format automation and common cryo racks.
  • 1.9 mL capacity.
  • Medical-grade cryogenic polypropylene (USP Class-6).
  • Three-code-in-one identification: 2D bottom barcode and 1D side barcode.
  • SBS-format external thread; caps supplied separated.
  • Electron beam sterilized, SAL = 10^-6.
  • Compatible with Brooks System and NEST racks.
  • Suitable for use at liquid nitrogen temperatures (-196 °C).
  • Packaged 48 per pack, 480 per case.