Learn More
Chemglass Life Sciences Cryogenic Vials, 4.0 mL, 3D Barcode, External Thread, Caps On, SBS Format
SDP
Catalog No. 502733353
Encompass_Preferred
Cryogenic vials with 4.0 mL capacity designed for SBS-format automated handling. These vials include a 3D barcode for reliable sample identification, external threads, and caps attached; they are supplied sterile and packaged for laboratory storage and automation.
  • 4.0 mL capacity.
  • 3D barcode for sample identification (digital code, 1D barcode, QR code).
  • External threaded vial with caps attached.
  • Sterile; electron beam sterilized, SAL = 10^-6.
  • Packaged 48 per pack, 480 per case.
  • SBS-format compatible for automated handling.
Catalog No. 50-273-3353 Supplier Chemglass Life Sciences Supplier No. CGN476502
Cryogenic vials with 4.0 mL capacity designed for SBS-format automated handling. These vials include a 3D barcode for reliable sample identification, external threads, and caps attached; they are supplied sterile and packaged for laboratory storage and automation.
  • 4.0 mL capacity.
  • 3D barcode for sample identification (digital code, 1D barcode, QR code).
  • External threaded vial with caps attached.
  • Sterile; electron beam sterilized, SAL = 10^-6.
  • Packaged 48 per pack, 480 per case.
  • SBS-format compatible for automated handling.