Learn More
USA Scientific Inc 1.0 mL Internal Thread Self-Standing ArcticIce Cryogenic Tube
Catalog No. 50352871
Encompass
1.0 mL ArcticIce cryogenic tube, internal thread, self-standing, internal V-bottom, 50/pack, sterile
Catalog No. 50-352-871 Supplier USA Scientific Inc Supplier No. 14107410
1.0 mL ArcticIce cryogenic tube, internal thread, self-standing, internal V-bottom, 50/pack, sterile