Learn More
USA Scientific Inc 1.0 mL External Thread Self-Standing ArcticIce Cryogenic Tube
Catalog No. 50352876
Encompass
1.0 mL ArcticIce cryogenic tube, external thread, self-standing, internal V-bottom, 50/pack, sterile
Catalog No. 50-352-876 Supplier USA Scientific Inc Supplier No. 14107310
1.0 mL ArcticIce cryogenic tube, external thread, self-standing, internal V-bottom, 50/pack, sterile