Learn More
USA Scientific Inc 1.8 mL Internal Thread Self-Standing ArcticIce Cryogenic Tube
Catalog No. 50352872
Encompass
1.8 mL ArcticIce cryogenic tube, internal thread, self-standing, internal U-bottom, 50/pack, sterile
Catalog No. 50-352-872 Supplier USA Scientific Inc Supplier No. 14187410
1.8 mL ArcticIce cryogenic tube, internal thread, self-standing, internal U-bottom, 50/pack, sterile