Learn More
USA Scientific Inc 1.8 mL Internal Thread Round Bottom ArcticIce Cryogenic Tube
Catalog No. 50352868
Encompass
1.8 mL ArcticIce cryogenic tube, internal thread, round bottom base, internal U-bottom, 50/pack, sterile
Catalog No. 50-352-868 Supplier USA Scientific Inc Supplier No. 14188410
1.8 mL ArcticIce cryogenic tube, internal thread, round bottom base, internal U-bottom, 50/pack, sterile