Learn More
USA Scientific Inc 1.8 mL External Thread Self-Standing ArcticIce Cryogenic Tube
Catalog No. 50352877
Encompass
1.8 mL ArcticIce cryogenic tube, external thread, self-standing, internal U-bottom, 50/pack, sterile
Catalog No. 50-352-877 Supplier USA Scientific Inc Supplier No. 14187310
1.8 mL ArcticIce cryogenic tube, external thread, self-standing, internal U-bottom, 50/pack, sterile