Learn More
USA Scientific Inc 3.6 mL Internal Thread Self-Standing ArcticIce Cryogenic Tube
Catalog No. 50352873
Encompass
3.6 mL ArcticIce cryogenic tube, internal thread, self-standing, internal U-bottom, 50/pack, sterile
Catalog No. 50-352-873 Supplier USA Scientific Inc Supplier No. 14367410
3.6 mL ArcticIce cryogenic tube, internal thread, self-standing, internal U-bottom, 50/pack, sterile