Learn More
USA Scientific Inc 4.5 mL Internal Thread Self-Standing ArcticIce Cryogenic Tube
Catalog No. 50352874
Encompass
4.5 mL ArcticIce cryogenic tube, internal thread, self-standing, internal U-bottom, 50/pack, sterile
Catalog No. 50-352-874 Supplier USA Scientific Inc Supplier No. 14457410
4.5 mL ArcticIce cryogenic tube, internal thread, self-standing, internal U-bottom, 50/pack, sterile