Learn More
Cryoguard Corporation CRYOGUARD M-40 10/PK
SDP
Catalog No. NC9773818
Encompass
Indicators Are Sealed In A Polycarbonate Plastic Housing Approximately 1 in. Long And 0.5 in. In Diameter. The Indicator Housing Contains Eyelets For Wiring It To The Monitored Material
Catalog No. NC9773818 Supplier Cryoguard Corporation Supplier No. M4010PK
Indicators Are Sealed In A Polycarbonate Plastic Housing Approximately 1 in. Long And 0.5 in. In Diameter. The Indicator Housing Contains Eyelets For Wiring It To The Monitored Material