Learn More
TWD Scientific LLC CRYOKING 1 INCH 100 WELL BLUE
SDP
Catalog No. NC1496766 Shop All TWD Scientific LLC Products
Encompass_Preferred
CryoKING 1 INCH 100 WELL BLUEPOLYCARBONATE SCAN-READYAUTOCLAVABLE CRYOGENIC STORAGE BOX(DIMENSIONS: 133x133x36mm). BOXES HAVEMARKED NUMERIC GRID FOR IDENTIFICATIONPURPOSES. 6 BOXES/PACK, 2 PACKS/CASE,PRICE/CASE
Catalog No. NC1496766 Supplier TWD Scientific LLC Supplier No. 980113
CryoKING 1 INCH 100 WELL BLUEPOLYCARBONATE SCAN-READYAUTOCLAVABLE CRYOGENIC STORAGE BOX(DIMENSIONS: 133x133x36mm). BOXES HAVEMARKED NUMERIC GRID FOR IDENTIFICATIONPURPOSES. 6 BOXES/PACK, 2 PACKS/CASE,PRICE/CASE