Learn More
GA International Cryogenic Multi Color Labels for 1.5 mL Microtubes 1.02" x 0.4" / 25.9mm x 10.2mm Rectangle 15 assorted colors From -196°C to +110°C (-321°F to +230°F)
Catalog No. NC0894629
Encompass_Preferred
Ideal for the identification of vials, microtubes, plastic bags, and other containers used in deep-freeze storage and color-coding in harsh laboratory environments. These labels are designed for reliable performance in extreme cold conditions, including liquid nitrogen exposure.They write easily with permanent markers—recommended options include MP-1, F-1, and SM-1.
Catalog No. NC0894629 Supplier GA International Supplier No. JTA2610A
Ideal for the identification of vials, microtubes, plastic bags, and other containers used in deep-freeze storage and color-coding in harsh laboratory environments. These labels are designed for reliable performance in extreme cold conditions, including liquid nitrogen exposure.They write easily with permanent markers—recommended options include MP-1, F-1, and SM-1.