Learn More
Electron Microscopy Sciences Cryo-Clear™ Laser Labels, 33 mm x 13 mm, Clear
SDP
Catalog No. 501900026
Encompass_Preferred
Clear cryogenic storage labels allow the vial contents beneath labels remain visible. The labels are temperature-resistant from -196 degree C to 70 degree C and remain fully adhered down to -196 degree C (liquid and vapor phase nitrogen). Chemically inert the labels resist most organic solvents and caustic agents and will adhere to plastics, glass and metals without cracking, peeling or degrading.
Label sheets are specially top-coated to accept laser printing and other markings. Heat-resistant, stay-flat sheets will not jam in laser printers.
Made with a flexible, vinyl label material and permanent acrylic adhesive. This unique combination expands and contracts together as temperatures fluctuate resulting in a strong and sustained adhesive bond.
Size: 33 mm x 13 mm
Color: Clear
Use: 1.5 ml tubes
Catalog No. 50-190-0026 Supplier Electron Microscopy Sciences Supplier No. 77575C
Clear cryogenic storage labels allow the vial contents beneath labels remain visible. The labels are temperature-resistant from -196 degree C to 70 degree C and remain fully adhered down to -196 degree C (liquid and vapor phase nitrogen). Chemically inert the labels resist most organic solvents and caustic agents and will adhere to plastics, glass and metals without cracking, peeling or degrading.
Label sheets are specially top-coated to accept laser printing and other markings. Heat-resistant, stay-flat sheets will not jam in laser printers.
Made with a flexible, vinyl label material and permanent acrylic adhesive. This unique combination expands and contracts together as temperatures fluctuate resulting in a strong and sustained adhesive bond.
Size: 33 mm x 13 mm
Color: Clear
Use: 1.5 ml tubes