Learn More
Electronic Imaging Materials Inc 667 Vial CryoLabel 1.00 inch x 0.75 inch, White, Perforated, 5K labels per roll, 3 inch core, 4 ROLL TIER
SDP
Catalog No. NC0878884
Encompass_Preferred
1.00 x 0.75 inch CryoLabel will handle extremely cold storage temperatures found in genetics, virology, microbiology and related fields. Conformable—works well on tight diameter test tubes. Resistant to moisture, smudging, and tearing, plus it holds up to alcohol and other chemicals. Designed for thermal transfer and durable UV inkjet printing, allowing you to color-code labels. Scratch and smear resistant when printed with T68 premium resin ribbon.
Catalog No. NC0878884 Supplier Electronic Imaging Materials Inc Supplier No. 6671000751 ROLL TIER
1.00 x 0.75 inch CryoLabel will handle extremely cold storage temperatures found in genetics, virology, microbiology and related fields. Conformable—works well on tight diameter test tubes. Resistant to moisture, smudging, and tearing, plus it holds up to alcohol and other chemicals. Designed for thermal transfer and durable UV inkjet printing, allowing you to color-code labels. Scratch and smear resistant when printed with T68 premium resin ribbon.