Learn More
Cayman Chemical Cryolys EvolutIon 1ea
SDP
Catalog No. 502979609
Encompass
A cell-permeable STAT3 activator (EC50 2 nM) that is more than 28000-fold selective for STAT3 over STAT1 and NF-KB increases the expression of BCL3 a known STAT3-dependent oncogene and enhances STAT3 activation by the pro-inflammatory cytokine IL-6
Catalog No. 50-297-9609 Supplier Cayman Chemical Supplier No. 169021 EA
A cell-permeable STAT3 activator (EC50 2 nM) that is more than 28000-fold selective for STAT3 over STAT1 and NF-KB increases the expression of BCL3 a known STAT3-dependent oncogene and enhances STAT3 activation by the pro-inflammatory cytokine IL-6