Learn More
Bertin Corp Cryolys Evolution
SDP
Catalog No. 501542928
Encompass_Preferred
Cryolys Evolution is the high-end cooling system specially developed for Precellys Evolution. Its innovative patented technology allows a very efficient cooling of the grinding area using dry ice and ambient air. Thanks to the exclusive software extension for Precellys Evolution, the temperature is set, controlled and maintained during all the process ensuring the protection of thermosensitive molecules such as enzymes, RNA or metabolites.
Catalog No. 50-154-2928 Supplier Bertin Corp Supplier No. P000671CLYS2A.0
Add to Cart
Add to Cart
Request a Quote
Cryolys Evolution is the high-end cooling system specially developed for Precellys Evolution. Its innovative patented technology allows a very efficient cooling of the grinding area using dry ice and ambient air. Thanks to the exclusive software extension for Precellys Evolution, the temperature is set, controlled and maintained during all the process ensuring the protection of thermosensitive molecules such as enzymes, RNA or metabolites.