Learn More
Cayman Chemical Cryolys EvolutIon EXended Wa
SDP
Catalog No. 502967485
Encompass
A histamine H1 receptor antagonist (Ki 0.98 nM for the human receptor) selective for histamine H1 over H3 and H4 receptors (Kis 100 and 77.6 UM respectively for the human receptors) binds mAChRs (Ki 22 nM) inhibits histamine-induced paw edema and acetic acid-induced writhing in mice (ED50s 5.9 and 11.8 mg/kg respectively) decreases the number of motion-induced vomiting episodes in S. murinus at 32 mg/kg
Catalog No. 50-296-7485 Supplier Cayman Chemical Supplier No. 173111 EA
A histamine H1 receptor antagonist (Ki 0.98 nM for the human receptor) selective for histamine H1 over H3 and H4 receptors (Kis 100 and 77.6 UM respectively for the human receptors) binds mAChRs (Ki 22 nM) inhibits histamine-induced paw edema and acetic acid-induced writhing in mice (ED50s 5.9 and 11.8 mg/kg respectively) decreases the number of motion-induced vomiting episodes in S. murinus at 32 mg/kg