Learn More
Cayman Chemical Cryolys Thermometer 1ea
SDP
Catalog No. 502984849
Encompass
A mitochondrial protonophore uncoupler that does not depolarize the plasma membrane and protects mice from acute renal ischemic-reperfusion injury
Catalog No. 50-298-4849 Supplier Cayman Chemical Supplier No. 205881 EA
A mitochondrial protonophore uncoupler that does not depolarize the plasma membrane and protects mice from acute renal ischemic-reperfusion injury