Learn More
Thermo Scientific™ CryoMed™ Bag Freezing Presses
Catalog No. 13262820
Change view
Click to view available options
Type:
Delmed 2030-2
Gambro DF-200
Gambro DF-700
Nexell R4R9955 (Fenwal 4R5462)
Holds:
Bags (2)
Bags (4)
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Holds
13-262-820 Delmed 2030-2 Bags (4)
13-262-821 Gambro DF-200 Bags (4)
13-262-822 Gambro DF-700 Bags (2)
11-679-125 Nexell R4R9955 (Fenwal 4R5462) Bags (2)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13-262-820 Supplier Thermo Scientific™ Supplier No. 4000317
Add to Cart
Add to Cart
Perform controlled-rate freezing applications with a controlled-rate freezer and these CryoMed bag freezing presses.
Perform controlled-rate freezing applications with a controlled-rate freezer and these Thermo Scientific™ CryoMed™ Bag Freezing Presses.
Bag presses for Nexell, Fenwal, Gambro and Delmed. Bag capacities 2 to 4 bags.
TRUSTED_SUSTAINABILITY
Specifications
Type Delmed 2030-2
For Use With (Equipment) CryoMed 7452, 7453, 7473, 7454, 7455, 7474, 7458
Holds Bags (4)