Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CryoMed™ Controlled-Rate Freezer, General Purpose
GSA_VA

Catalog No. TSCM17PA
Change view
Click to view available options
Capacity (Metric):
17 L
34 L
48 L
Electrical Requirements:
120 V, 60Hz
Model:
General Purpose with Printer
General Purpose without Printer
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Electrical Requirements Model
TSCM17PA 17 L 120 V, 60Hz General Purpose with Printer
TSCM34PA 34 L 120 V, 60Hz General Purpose with Printer
TSCM48PA 48 L 120 V, 60Hz General Purpose with Printer
TSCM17XA 17 L 120 V, 60Hz General Purpose without Printer
TSCM34XA 34 L 120 V, 60Hz General Purpose without Printer
TSCM48XA 48 L 120 V, 60Hz General Purpose without Printer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. TSCM17PA Supplier Thermo Scientific™ Supplier No. TSCM17PA
Get Thermo Scientific Accessories with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Thermo Scientific CryoMed Controlled-Rate Freezers (CRF) are designed with reliability, protection, and data traceability in mind for your sample preparation needs.

For customers in research, cell and gene therapy, vaccine production, and biobanking, the CryoMed Controlled-Rate Freezer (CRF) provides precise, repeatable freezing results that protect samples from intracellular freezing. The CryoMed CRF features enhanced data traceability via a touch screen user interface and offers customizable freezing profiles while supporting 21 CFR part 11 and GMP needs.

Thermo Scientific CryoMed Controlled-Rate Freezers (CRF) are designed with reliability, protection, and data traceability in mind for your sample preparation and GMP production needs.

User-friendly operation with enhanced data traceability

  • Intuitive touchscreen display allows for easy set-up, operation, and review of a freezing run
  • Six pre-set freezing profiles and space for up to 14 user-defined, 'custom' freeze profiles
  • Integrated touchscreen UI logs usage and events to support 21 CFR part 11 requirements
  • USB data export of .pdf file run logs support 21 CFR part 11
  • User security: 3 levels of user accessibility
  • 'Run last' feature allows the same profile to be run consecutively with the touch of a button
  • PC interface software provides remote operation capabilities, run review, and custom profile creation

Real-time run monitoring for sample protection

  • Chamber and sample temperatures are monitored by Type T thermocouples, eliminating lag time and providing 'real time' responsiveness
  • Standard alarms to alert users of thermocouple failures, heater malfunction, high/low temperature limits, temperature tracking, power failure, and completion of run
  • Remote alarm contacts
  • Optional thermal printer

Reliable temperature performance for high throughput usage

  • Dual solenoid valves are designed to balance LN2 injection volume for precise temperature control and accelerated freezing
  • Preventative maintenance indicator for LN2 solenoid replacement limits downtime
  • Temperature control and uniformity achieved via an air-handling and liquid nitrogen injection ring system
  • Compatible with VHP cleaning techniques
  • Environmentally friendly insulating foam
  • Type 304 stainless steel with exterior powder-coat finish

Warranty varies by region. Check with your local representative for more information

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Type Controlled-rate freezer
Capacity (English) 0.6 cu. ft.
Capacity (Metric) 17 L
Temperature Range -180°C to +50°C
No. of Doors 1
Cabinet Material Stainless Steel
Exterior Finish Powder-coated Steel
Certifications/Compliance cULus
Style Benchtop
Display Touchscreen User Interface
Warranty Warranty varies by region. Contact your local sales representative for more information
Model General Purpose with Printer
Sample Loading Front-loading
Dimensions (DxWxH) Exterior 24.3 x 37.3 x 21.7 in. (61 x 94.7 x 55.1 cm)
Dimensions (D x W x H) Interior 13 x 7 x 12 in. (33 x 17.8 x 30.5 cm)
Monitoring Options Alarms, Remote Alarm Contacts
Data Outputs USB data export
On-Board Datalogging Yes
PC Programmability PC Interface Software Supported
Connectivity Thermo Fisher Connect compatible
Amperage 12 A
Plug Type NEMA 5-15 P
Voltage 120 V
Electrical Requirements 120 V, 60Hz
Vial Capacity <380
Compressor Air-cooled
Weight (English) 154 lb.
Weight (Metric) 69.8 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.