Learn More
Andwin Scientific CRYOMOLD 15X15X5
SDP
Catalog No. NC9464347
Encompass_Preferred
The Cryomold molds produce uniformly shaped, flat-surface specimen block when used with O.C.T. (optimum cutting temperature) compound, which is sold separately, for frozen sectioning. This can fit into Cryobar freezing wells of the Tissue Tek II microtome/cryostat with the use of an adapter. This product measures 15 mm x 15 mm x 15 mm.
Catalog No. NC9464347 Supplier Andwin Scientific Supplier No. 25608924
The Cryomold molds produce uniformly shaped, flat-surface specimen block when used with O.C.T. (optimum cutting temperature) compound, which is sold separately, for frozen sectioning. This can fit into Cryobar freezing wells of the Tissue Tek II microtome/cryostat with the use of an adapter. This product measures 15 mm x 15 mm x 15 mm.