Learn More
Andwin Scientific CRYOMOLDS PK/100CRYOMOLDS
SDP
Catalog No. NC9643511
Encompass_Preferred
The Cryomold molds produce uniformly shaped, flat-surface specimen block when used with O.C.T. (optimum cutting temperature) compound, which is sold separately, for frozen sectioning. This can fit into Cryobar freezing wells of the Tissue Tek II microtome/cryostat with the use of an adapter. This product measures 25mm x 20mm.
Catalog No. NC9643511 Supplier Andwin Scientific Supplier No. 25608916
The Cryomold molds produce uniformly shaped, flat-surface specimen block when used with O.C.T. (optimum cutting temperature) compound, which is sold separately, for frozen sectioning. This can fit into Cryobar freezing wells of the Tissue Tek II microtome/cryostat with the use of an adapter. This product measures 25mm x 20mm.