Learn More
ILEX LIFE SCIENCES LLC CRYOPAN II SFM 6 X 100 ML
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0793100X6
Encompass_Preferred
PAN-Biotech Cryopan II, Chemically-Defined Serum-Free and Protein-Free Freezing Medium (6 x 100 ml), Cryopreservation Media. DESCRIPTION: PAN-Biotech Cryopan II is a chemically-defined, serum-free and protein-free freezing medium that is suitable for the cryopreservation of various cell types, including both primary and established cell lines. Its chemically defined composition, free from human or animal components, ensures optimal cell preservation. Cryopan II can be used for freezing adherent or suspension cells, making it a versatile option for the cryopreservation of animal and human cells across different cell types and culture conditions. COMPOSITION: Cryopan II consists of a chemically defined and optimized mixture of salts, sugar, 10% DMSO and additional antifreeze-substances. It contains no animal and human components. LIQUID/POWDER: Liquid. STERILE: Yes. STORAGE TEMP: -20°C. MANUFACTURER: PAN-Biotech GmbH (Germany).
Catalog No. 50-273-3704