Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Cryopreservation Medium with FBS and DMSO, 50 mL
SDP

Catalog No. 502209756
Encompass_Preferred

Cryopreservation Medium is a ready-to-use medium for the preservation of cells as seed stock or for future investigative study. Cryopreservation allows you to preserve and protect your cell line stock(s) from over culturing that can lead to unwanted changes within cellular properties or lead to senescence.

Ready-to-use
Optimizes cell growth and cell viability after thawing
Reduces potential for transmission of infectious diseases
Contains Fetal Bovine Serum Calf Serum and DMSO

Storage Condition: -10 °C to -20 °C
Shipping Condition: on dry ice
Performance Tested
pH: 6.1 ± 0.2
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-220-9756 Supplier Quality Biological Inc Supplier No. 118298051
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cryopreservation Medium is a ready-to-use medium for the preservation of cells as seed stock or for future investigative study. Cryopreservation allows you to preserve and protect your cell line stock(s) from over culturing that can lead to unwanted changes within cellular properties or lead to senescence.

Ready-to-use
Optimizes cell growth and cell viability after thawing
Reduces potential for transmission of infectious diseases
Contains Fetal Bovine Serum Calf Serum and DMSO

Storage Condition: -10 °C to -20 °C
Shipping Condition: on dry ice
Performance Tested
pH: 6.1 ± 0.2
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.