Learn More
Lonza Walkersville Mouse C57BL/6 Hepatocytes, Cryopreserved, Plateable
Catalog No. NC1845037
Encompass
Cryopreserved C57BL/6 Mouse Hepatocytes, Plateable are specifically pre tested for metabolic activity of general Phase I and Phase II metabolism in suspension.They are tested for healthy morphology lasting 72 hours in monlayer sandwich culture.Applications:Drug metabolism in plated monolayer formatSpecies-species comparisonsGenomicsContent:1 x Cryopreserved ampule of C57BL/6 Mouse Hepatocytes (plateable format) containing > 5 million viable cells
Catalog No. NC1845037 Supplier Lonza Walkersville Supplier No. MBCP01
Cryopreserved C57BL/6 Mouse Hepatocytes, Plateable are specifically pre tested for metabolic activity of general Phase I and Phase II metabolism in suspension.They are tested for healthy morphology lasting 72 hours in monlayer sandwich culture.Applications:Drug metabolism in plated monolayer formatSpecies-species comparisonsGenomicsContent:1 x Cryopreserved ampule of C57BL/6 Mouse Hepatocytes (plateable format) containing > 5 million viable cells