Learn More
Cell Applications Inc 3-D Skin Model Total Kit
SDP
Supplier:  Cell Applications Inc 102K3D
Encompass_Preferred
3-D Skin Model Total Kit **(102-3D) Cryopreserved HEK, 1 ampoule (131-3D-100) 3-D Keratinocyte Growth Media, 100ml (090mK) Subculture Reagent Mini-KitHBSS, 10ml; Trypsin/EDTA, 10ml; Trypsin Neutralization Solution, 10ml(013-14) Millicell PCF inserts (014-01) Fixative 1 (015-01) Crystal Violet (131D-3D-150) 3-D Airway Differentiation Media, 150ml (016-01) Fixative 2
Catalog No. 50-195-9046