Learn More
Cell Applications Inc Rat Striatal Neuron Total Kit
SDP
Catalog No. 501959231
Encompass
Rat Striatal Neuron Total Kit:(R8812N-10) Cryopreserved (RStN), 1 vial(R817-100) Rat Neuron Culture Media, 100ml(027-05) Neuron Coating Solution I, 5ml(R817P-10) Neuron Plating Solution, 10ml
Catalog No. 50-195-9231 Supplier Cell Applications Inc Supplier No. R8812NK10
Rat Striatal Neuron Total Kit:(R8812N-10) Cryopreserved (RStN), 1 vial(R817-100) Rat Neuron Culture Media, 100ml(027-05) Neuron Coating Solution I, 5ml(R817P-10) Neuron Plating Solution, 10ml