Learn More
Cell Applications Inc Human Neural Stem Cell Total Kit, fetal
SDP
Catalog No. 501959190
Encompass
Human Neural Stem Cell Total Kit, fetal(HS820-20f) Cryopreserved (HNSC), fetal(813-250) Neural Stem Cell Growth Media, 250ml(076-20) Neural Stem Cell Dissociation Solution, 20ml(2) 10cm non-tissue culture dishes
Catalog No. 50-195-9190 Supplier Cell Applications Inc Supplier No. HS820K20F
Human Neural Stem Cell Total Kit, fetal(HS820-20f) Cryopreserved (HNSC), fetal(813-250) Neural Stem Cell Growth Media, 250ml(076-20) Neural Stem Cell Dissociation Solution, 20ml(2) 10cm non-tissue culture dishes