Learn More
Cell Applications Inc Human Jejunum Epithelial Cells Total Kit, adult
SDP
Supplier:  Cell Applications Inc 732JEK10A
Encompass_Preferred
Human Jejunum Epithelial Cells Total Kit, adult:(732Je-10a) Cryopreserved (HJeEpC) 1 vial, adult(716DCMK-50) GI Epithelial Cell Defined Culture Media, 50ml(025-05) Coating Solution for GI Epithelial Cells(716T-20) Thawing Solution for GI Epithelial Cells
Catalog No. 50-195-9146