Learn More
Cell Applications Inc Human Colonic Epithelial Cells Total Kit, fetal
SDP
Supplier:  Cell Applications Inc 732CNK10F
Encompass_Preferred
Human Colonic Epithelial Cells Total Kit, fetal:(732Cn-10F) Cryopreserved (HCnEpC) 1 vial, fetal(716DCMK-50) GI Epithelial Cell Defined Culture Media, 50ml(025-05) Coating Solution for GI Epithelial Cells(716T-20) Thawing Solution for GI Epithelial Cells
Catalog No. 50-195-9134