Learn More
Cell Applications Inc Human Colonic Epithelial Cells Total Kit, adult
SDP
Supplier:  Cell Applications Inc 732CNK10A
Encompass_Preferred
Human Colonic Epithelial Cells Total Kit, adult:(732Cn-10A) Cryopreserved (HCnEpC) 1 vial, adult(716DCMK-50) GI Epithelial Cell Defined Culture Media, 50ml(025-05) Coating Solution for GI Epithelial Cells(716T-20) Thawing Solution for GI Epithelial Cells
Catalog No. 50-195-9133