Learn More
Cell Applications Inc Mouse Cortical Neurons Total Kit
SDP
Catalog No. 501959197
Encompass
Mouse Cortical Neurons Total Kit:(M882N-20) Cryopreserved (MCoN), 1 vial(M817K-100) Mouse Neuron Culture Media Kit, 100ml(M817P-10) Mouse Neuron Plating Media, 10ml(027-05) Neuron Coating Solution I, 5ml
Catalog No. 50-195-9197 Supplier Cell Applications Inc Supplier No. M882NK20
Mouse Cortical Neurons Total Kit:(M882N-20) Cryopreserved (MCoN), 1 vial(M817K-100) Mouse Neuron Culture Media Kit, 100ml(M817P-10) Mouse Neuron Plating Media, 10ml(027-05) Neuron Coating Solution I, 5ml