Learn More
Lonza Walkersville AoSMC – Human Aortic Smooth Muscle Cells
Catalog No. NC9679464
Encompass
Human Vascular Smooth Muscle Cells come from multiple tissue types including Aorta (AoSMC), Coronary Artery (CASMC), Pulmonary Artery (PASMC), and Umbilical Artery (UASMC). Cryopreserved cells are shipped in 3rd passage.Clonetics Human Aortic Smooth Muscle Cells are guaranteed for 15 population doublings and to stain positive for alpha smooth muscle actin and negative for von Willebrand Factor VIII after differentiation. All cells test negative for mycoplasma, bacteria, yeast and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis (CoA) is provided for each cell lot purchased.Applications:StrokeDiabetes Vascular research Artherosclerosis HypertensionContent:1 x Cryopreserved ampule of AoSMC containing ≥ 500,000 cells/vial
This item is not returnable. View return policy
Catalog No. NC9679464 Supplier Lonza Walkersville Supplier No. CC2571
Add to Cart
Add to Cart
Human Vascular Smooth Muscle Cells come from multiple tissue types including Aorta (AoSMC), Coronary Artery (CASMC), Pulmonary Artery (PASMC), and Umbilical Artery (UASMC). Cryopreserved cells are shipped in 3rd passage.Clonetics Human Aortic Smooth Muscle Cells are guaranteed for 15 population doublings and to stain positive for alpha smooth muscle actin and negative for von Willebrand Factor VIII after differentiation. All cells test negative for mycoplasma, bacteria, yeast and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis (CoA) is provided for each cell lot purchased.Applications:StrokeDiabetes Vascular research Artherosclerosis HypertensionContent:1 x Cryopreserved ampule of AoSMC containing ≥ 500,000 cells/vial