Learn More
Andwin Scientific CRYOPRO STORAGE BOXES AND DIV
SDP
Catalog No. NC9888486
Encompass_Preferred
The CryoPro Fiberboard cardboard cell divider feature numbered grids that allow users to have easy identification and organization of items. This product measures 13.2 x 13.2 x 7.6 cm (51/5 x 51/5 x 3"). It is made of durable fiberboard with a protective, moisture-repellant coating.
Catalog No. NC9888486 Supplier Andwin Scientific Supplier No. 82021116
The CryoPro Fiberboard cardboard cell divider feature numbered grids that allow users to have easy identification and organization of items. This product measures 13.2 x 13.2 x 7.6 cm (51/5 x 51/5 x 3"). It is made of durable fiberboard with a protective, moisture-repellant coating.