Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ad Infinitum™ CryoProtectPURE™ Cryopreservation Media

Catalog No. CPP100025 Shop All Ad Infinitum Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CPP100025 25 mL
CPP100100 100 mL
CPP100500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CPP100025 Supplier Ad Infinitum™ Supplier No. CPP100025
Add to Cart
Add to Cart

Ad Infinitum™ CryoProtectPURE™ Cryopreservation Media is a DMSO-Free and Serum-Free cryopreservation media for cell and tissue preservation.

  • Increases the cell viability during freeze and post-thaw, producing healthier cells with no antigenicity concern or toxic vitrification.
  • Serum-Free
  • Pre-mixed
  • Ready-to-use
  • 0.2 μm filtered
  • Suitable for a wide variety of cells
  • Shipped at ambient temperature
  • For manufacturing and/or research use only, not for diagnostic or therapeutic use
  • Manufactured at a cGMP-compliant facility registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards

Specifications

Product Type Cryopreservation Media
For Use With (Application) Manufacturing and/or research use only, not for diagnostic or therapeutic use
Quantity 25 mL
Certifications/Compliance ISO 13485 standards
Content And Storage 2 to 8°C
Description Pre-mixed and ready-to-use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.