Learn More
Sarstedt Inc CRYOPURE,1.2ML,EXT CAP/PK500
Catalog No. 50873790 Shop All Sarstedt Inc. Products
Encompass_Preferred
Tube, Cyrogenic; CyroPure; Sarstedt; Vol.: 1.0mL; With external thread; Sterile; Red cap; Interlocking base enables single-handed operation in CryoRack 40 and most cryogenic racks; Fit standard cryo boxes and aluminium holders; 500/pk.
Catalog No. 50-873-790 Supplier Sarstedt Inc Supplier No. 72.377.002
Tube, Cyrogenic; CyroPure; Sarstedt; Vol.: 1.0mL; With external thread; Sterile; Red cap; Interlocking base enables single-handed operation in CryoRack 40 and most cryogenic racks; Fit standard cryo boxes and aluminium holders; 500/pk.