Learn More
Sarstedt Inc Tube, Cryogenic, 1.2mL, External Thread, Green Cap, 500/Pack
Catalog No. 50873792 Shop All Sarstedt Inc. Products
Encompass_Preferred
Tube, Cyrogenic; CyroPure; Sarstedt; Vol.: 1.0mL; With external thread; Sterile; Green cap; Interlocking base enables single-handed operation in CryoRack 40 and most cryogenic racks; Fit standard cryo boxes and aluminium holders; 500/pk.
Catalog No. 50-873-792 Supplier Sarstedt Inc Supplier No. 72.377.005
Tube, Cyrogenic; CyroPure; Sarstedt; Vol.: 1.0mL; With external thread; Sterile; Green cap; Interlocking base enables single-handed operation in CryoRack 40 and most cryogenic racks; Fit standard cryo boxes and aluminium holders; 500/pk.