Learn More
Sarstedt Inc CRYOPURE 1.2ML EXT YELLO/PK500
Catalog No. 50873791 Shop All Sarstedt Inc. Products
Encompass_Preferred
Tube, Cyrogenic; CyroPure; Sarstedt; Vol.: 1.0mL; With external thread; Sterile; Yellow cap; Interlocking base enables single-handed operation in CryoRack 40 and most cryogenic racks; Fit standard cryo boxes and aluminium holders; 500/pk.
Catalog No. 50-873-791 Supplier Sarstedt Inc Supplier No. 72.377.004
Tube, Cyrogenic; CyroPure; Sarstedt; Vol.: 1.0mL; With external thread; Sterile; Yellow cap; Interlocking base enables single-handed operation in CryoRack 40 and most cryogenic racks; Fit standard cryo boxes and aluminium holders; 500/pk.