Learn More
Sarstedt Inc CRYOPURE,2ML,INT,RED CAP/PK500
Catalog No. 50873800 Shop All Sarstedt Inc. Products
Encompass_Preferred
Tube, Cyrogenic; CyroPure; Sarstedt; Vol.: 1.6mL; With internal thread; Sterile; Red cap; Interlocking base enables single-handed operation in CryoRack 40 and most cryogenic racks; Fit standard cryo boxes and aluminium holders; 500/pk.
Catalog No. 50-873-800 Supplier Sarstedt Inc Supplier No. 72.380.002
Tube, Cyrogenic; CyroPure; Sarstedt; Vol.: 1.6mL; With internal thread; Sterile; Red cap; Interlocking base enables single-handed operation in CryoRack 40 and most cryogenic racks; Fit standard cryo boxes and aluminium holders; 500/pk.