Learn More
Sarstedt Inc Tube, Cryogenic, 2.0mL, Internal Thread, White Cap, 500/Pack
Catalog No. 50873799 Shop All Sarstedt Inc. Products
Encompass_Preferred
Tube, Cyrogenic; CyroPure; Sarstedt; Vol.: 1.6mL; With internal thread; Sterile; White cap; Interlocking base enables single-handed operation in CryoRack 40 and most cryogenic racks; Fit standard cryo boxes and aluminium holders; 500/pk.
Catalog No. 50-873-799 Supplier Sarstedt Inc Supplier No. 72.380
Tube, Cyrogenic; CyroPure; Sarstedt; Vol.: 1.6mL; With internal thread; Sterile; White cap; Interlocking base enables single-handed operation in CryoRack 40 and most cryogenic racks; Fit standard cryo boxes and aluminium holders; 500/pk.