Learn More
Sarstedt Inc CRYOPURE 5ML EXT YELLOW/PK500
Catalog No. 50873806 Shop All Sarstedt Inc. Products
Encompass_Preferred
Tube, Cyrogenic; CyroPure; Sarstedt; Vol.: 4.5mL; With external thread; Sterile; Yellow cap; Interlocking base enables single-handed operation in CryoRack 40 and most cryogenic racks; Fit standard cryo boxes and aluminium holders; 250/pk.
Catalog No. 50-873-806 Supplier Sarstedt Inc Supplier No. 72.383.004
Tube, Cyrogenic; CyroPure; Sarstedt; Vol.: 4.5mL; With external thread; Sterile; Yellow cap; Interlocking base enables single-handed operation in CryoRack 40 and most cryogenic racks; Fit standard cryo boxes and aluminium holders; 250/pk.