Learn More
Research Products International Corp Cryo-Safe Cooler with Clear Lid, Holds 0.5, 1.5, or 2.0ml Tubes, 12 Tube Capacity
SDP
Catalog No. 50476449
Encompass
This small cooler safely holds twelve 0.5, 1.5 or 2.0ml microcentrifuge tubes or cryogenic vials at a temperature below -15ºC for approximately 1 hour. Ideal for protecting valuable samples stored in a freezer with defrost cycle. Use included insert to hold 0.5ml microcentrifuge tubes. Store at -20ºC for 24 hours prior to use. Clear polycarbonate lid shows positions of tubes; keyed to correspond with positions printed on cooler. Molded side handles allow secure grip for transfer. Rubber feet prevent sliding on the lab bench and coolers are stackable.
WARNING  This product can expose you to chemicals including Bisphenol-A , which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-476-449 Supplier Research Products International Corp Supplier No. 142410
This small cooler safely holds twelve 0.5, 1.5 or 2.0ml microcentrifuge tubes or cryogenic vials at a temperature below -15ºC for approximately 1 hour. Ideal for protecting valuable samples stored in a freezer with defrost cycle. Use included insert to hold 0.5ml microcentrifuge tubes. Store at -20ºC for 24 hours prior to use. Clear polycarbonate lid shows positions of tubes; keyed to correspond with positions printed on cooler. Molded side handles allow secure grip for transfer. Rubber feet prevent sliding on the lab bench and coolers are stackable.
WARNING  This product can expose you to chemicals including Bisphenol-A , which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.