Learn More
Research Products International Corp Cryo-Safe Cooler with Gel filled Lid, Holds 0.5, 1.5, or 2.0 ml Tubes, 32 Tube Capacity
SDP
Catalog No. 50476450
Encompass
This cooler safely holds thirty-two 0.5, 1.5 or 2.0 ml microcentrifuge tubes or cryogenic vials at a temperature below -15ºC for approximately 1 1/2 hours. Use included insert to hold 0.5ml microcentrifuge tubes. Store at -20ºC for 24 hours prior to use. Ideal for protecting valuable samples stored in a freezer with defrost cycle. Polycarbonate cooler body is filled with a non-toxic gel which cannot contact tubes; ensures markings and labels on tubes are safe. Gel-filled polycarbonate lid extends temperature holding time; secured closed with a built-in wire carrying handle. .
WARNING  This product can expose you to chemicals including Bisphenol-A, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-476-450 Supplier Research Products International Corp Supplier No. 142412
This cooler safely holds thirty-two 0.5, 1.5 or 2.0 ml microcentrifuge tubes or cryogenic vials at a temperature below -15ºC for approximately 1 1/2 hours. Use included insert to hold 0.5ml microcentrifuge tubes. Store at -20ºC for 24 hours prior to use. Ideal for protecting valuable samples stored in a freezer with defrost cycle. Polycarbonate cooler body is filled with a non-toxic gel which cannot contact tubes; ensures markings and labels on tubes are safe. Gel-filled polycarbonate lid extends temperature holding time; secured closed with a built-in wire carrying handle. .
WARNING  This product can expose you to chemicals including Bisphenol-A, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.