Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ CryoSavers™ Brucella Broth with 10% Glycerol and Beads
SDP

Catalog No. 23001753 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Fill Volume:
1.0 to 1.4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fill Volume
23-001-753 1.0 to 1.4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-753 Supplier Hardy Diagnostics™ Supplier No. CS100B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recommended for the preservation of microorganisms by freezing. It is formulated using 10% glycerol.

  • High concentration of glycerol acts to prevent cell damage in the harsh environment of sub-zero temperatures
  • The cleaned, acid-washed beads are of porous nature allowing microorganisms to readily adhere to the bead surface
  • Offers a simple, safe, and convenient way to store organisms at ultra low temperatures for future use
  • Caps are ergonomically designed to feature large ridges for easy opening. Threads are located on the outside of the vial to reduce the chance of contamination
  • Available formulas include Skim Milk or Brucella Broth for the purpose of growth and long term viability of your organism
  • Ready-to-use media with cryo-preservatives saves you time and expense of preparing the media yourself
  • Each vial has a blank white area for written identification. Additionally, each box comes with a moisture-proof card to record the contents of individual vials

Specifications

Appearance Clear and Amber (20 Beads and Holes)
Broth Type Brucella
Cap Color Opaque
Certifications/Compliance ANAB Calibration ISO 17025, BSI-ISO 13485
Color Amber
Concentration 10% Glycerol
Content And Storage 2°C to 8°C
Additive Glycerol
Detects Microorganisms
Format Liquid, Broth Tube, Cryogenic Vial, Ready-to-use media
Manufacturing Origin United States
Media Type Brucella Broth
Product Type Broth Tube
Sterility Sterile
Fill Volume 1.0 to 1.4 mL
Formulation Pancreatic Digest of Casein (10.0gm), Peptic Digest of Animal Tissue (10.0gm), Sodium Chloride (5.0gm), Yeast Extract (2.0gm), Dextrose (1.0gm), Sodium Bisulfite (0.1gm), Glycerol (100.0ml); Final pH: 7.0 ± 0.3 at 25°C
For Use With (Application) Preservation of microorganisms by freezing
Includes Plastic Beads, Moisture-proof Card
Incubation Time Overnight
Material Polypropylene (vial, cap inserts), Plastic (beads)
No. per Pack 100
Packaging Type Box, Vials
Quantity 100 Vials
Protein Source Casein, Animal Tissue, Yeast
Manufacturing Quality ANAB Calibration ISO 17025, BSI-ISO 13485
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.