Learn More
Electron Microscopy Sciences Frozen Tissue Storage Container, 50/PK
SDP
Catalog No. 502090678
Encompass_Preferred
The CryoSette is designed for frozen tissue collection, transport and storage. The robust design ensures that tissue morphology will be well preserved. This 2.5 ml container features a wide-mouth opening and a high integrity screw closure. Designed with a flat bottom and straight sides for easy tissue removal. The specific shape allows its manipulation to be optimized, whether you are using gloves or not. The CryoSette efficiently stores tissue samples in -86 degree C ULT freezers or in the gas phase of liquid nitrogen in Dewar flasks at -196 degree C, but not in LN2 liquid phase. The screw closure and base are designed so they will not seize at low temperatures. Rapid visual 1¼ turn, even during repeated freeze/thaw cycles. The CryoSette offers two surfaces for handwriting or applying labels: an anterior surface and a larger space underneath the base. The anterior writing surface is improved for ink adherence. Tested at 95 kPa.
Catalog No. 50-209-0678 Supplier Electron Microscopy Sciences Supplier No. 6201401
The CryoSette is designed for frozen tissue collection, transport and storage. The robust design ensures that tissue morphology will be well preserved. This 2.5 ml container features a wide-mouth opening and a high integrity screw closure. Designed with a flat bottom and straight sides for easy tissue removal. The specific shape allows its manipulation to be optimized, whether you are using gloves or not. The CryoSette efficiently stores tissue samples in -86 degree C ULT freezers or in the gas phase of liquid nitrogen in Dewar flasks at -196 degree C, but not in LN2 liquid phase. The screw closure and base are designed so they will not seize at low temperatures. Rapid visual 1¼ turn, even during repeated freeze/thaw cycles. The CryoSette offers two surfaces for handwriting or applying labels: an anterior surface and a larger space underneath the base. The anterior writing surface is improved for ink adherence. Tested at 95 kPa.