Learn More
Electron Microscopy Sciences Storage Boxes, 10/PK
SDP
Catalog No. 502090686
Encompass_Preferred
CryoSette Storage boxes are designed to hold frozen tissue Cryosette containers. These innovative boxes can be stored in low-temperature freezers from -196 degrees celsium to 121 degrees celsius. They are suitable for cryogenic storage, but only in gas phase of liquid nitrogen. Made of polypropylene.
Catalog No. 50-209-0686 Supplier Electron Microscopy Sciences Supplier No. 6201601
CryoSette Storage boxes are designed to hold frozen tissue Cryosette containers. These innovative boxes can be stored in low-temperature freezers from -196 degrees celsium to 121 degrees celsius. They are suitable for cryogenic storage, but only in gas phase of liquid nitrogen. Made of polypropylene.