Learn More
Sigma Aldrich Fine Chemicals Biosciences CryoSOfreeTM DMSO free Cry
Catalog No. 502905215
Encompass
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry compared to the standard use of DMSO for cryopreservation of cell cultures. for more information see here.
Catalog No. 50-290-5215 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C9249100ML
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry compared to the standard use of DMSO for cryopreservation of cell cultures. for more information see here.