Learn More
Molecular Dimensions CRYOSOL
SDP
Catalog No. NC0837597
Encompass_Preferred
Protein Crystallography. CryoSol (33 x 1.5mL):an extended set of multicomponent solutions intended for crystal cryoprotection and soaking of hydrophobic ligands into crystals. Compliments CryoProtX.
Catalog No. NC0837597 Supplier Molecular Dimensions Supplier No. MD190
Protein Crystallography. CryoSol (33 x 1.5mL):an extended set of multicomponent solutions intended for crystal cryoprotection and soaking of hydrophobic ligands into crystals. Compliments CryoProtX.