Learn More
RWD LIFE SCIENCE INC Cryostat FS800 Semi-Automatic FREEZING MICROTOME
SDP
Catalog No. NC3404381
Encompass_Preferred
The RWD FS800 Semi-Automatic Cryostat is designed for microtomy, intraoperative frozen sections, frozen tissue preparation, and histopathology. FDA and ISO13485 certified, it offers stable performance with advanced temperature control, a precise cross rail, high-resolution motor, and aviation aluminum slider for consistent slice quality. Safety features include low-temperature UVC disinfection and a waste liquid management system with a full bottle alarm. The cryostat’s touch-sensitive screen integrates sample feeding, trimming, section switching, and parameter adjustments, while a visual pointer ensures precise positioning. Automatic sleep/wake-up functions conserve energy and streamline operation.
Catalog No. NC3404381 Supplier RWD LIFE SCIENCE INC Supplier No. FS800
Add to Cart
Add to Cart
Request a Quote
The RWD FS800 Semi-Automatic Cryostat is designed for microtomy, intraoperative frozen sections, frozen tissue preparation, and histopathology. FDA and ISO13485 certified, it offers stable performance with advanced temperature control, a precise cross rail, high-resolution motor, and aviation aluminum slider for consistent slice quality. Safety features include low-temperature UVC disinfection and a waste liquid management system with a full bottle alarm. The cryostat’s touch-sensitive screen integrates sample feeding, trimming, section switching, and parameter adjustments, while a visual pointer ensures precise positioning. Automatic sleep/wake-up functions conserve energy and streamline operation.